missing translation for 'onlineSavingsMsg'
Learn More

Kaempferide, TRC

Product Code. 17968324
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17968324 25 mg 25mg
17907164 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17968324 Supplier Toronto Research Chemicals Supplier No. TRCK10006025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-54-3
Molecular Formula C16 H12 O6
Molecular Weight (g/mol) 300.26
Synonym 4H-1-Benzopyran-4-one, 3,5,7-trihydroxy-2-(4-methoxyphenyl)-, Flavone, 3,5,7-trihydroxy-4'-methoxy- (7CI,8CI), Kaempferide (6CI), 3,5,7-Trihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 3,5,7-Trihydroxy-4'-methoxyflavone, 4'-Methoxykaempferol, 4'-Methylkaempferol, 4'-O-Methylkaempferol, 4'-O-Methylkaempherol, 5,7-Dihydroxy-4'-methoxyflavonol, Kaempferid, Kaempferol 4'-O-methyl ether, Kaempferol 4'-methyl ether, Kaempherol 4'-O-methyl ether, NSC 407294
IUPAC Name 3,5,7-trihydroxy-2-(4-methoxyphenyl)chromen-4-one
SMILES COc1ccc(cc1)C2=C(O)C(=O)c3c(O)cc(O)cc3O2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H12O6/c1-21-10-4-2-8(3-5-10)16-15(20)14(19)13-11(18)6-9(17)7-12(13)22-16/h2-7,17-18,20H,1H3
Formula Weight 300.0634
Chemical Name or Material Kaempferide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.