Learn More
Justrite™ Document Storage Boxes
Product Code. 10830990
Change view
Click to view available options
Dimensions (L x W x H):
1.62 x 8.75 x 11.5 in.
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
10830990 1.62 x 8.75 x 11.5 in. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10830990 Supplier Justrite™ Supplier No. 23303
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Description Medium top opening
Product Type Document Storage Box
Color Red
Material Polyethylene
Includes Label pack with 2 blank labels and labels printed SDS and instruction manual
For Use With (Application) Used to keep important papers readily available while protecting them from moisture and dirt
Certifications/Compliance OSHA
Dimensions (L x W x H) 1.62 x 8.75 x 11.5 in.
Height (English) 12.5 in.
Height (English) Interior 11.5 in.
Height (Metric) 31.8 cm
Height (Metric) Interior 29.2 cm
Length (English) 2.25 in.
Length (English) Interior 1.62 in.
Length (Metric) 5.7 cm
Length (Metric) Interior 4.1 cm
Width (English) 10.25 in.
Width (English) Interior 8.75 in.
Width (Metric) 26 cm
Width (Metric) Interior 22.2 cm