missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNJ16259685, MedChemExpress
Description
JNJ16259685 is a selective antagonist of mGlu1 receptor, and inhibits the synaptic activation of mGlu1 in a concentration-dependent manner with IC50 of 19 nM.
Specifications
Specifications
| Chemical Name or Material | JNJ16259685 |
| Physical Form | Solid |
| CAS | 409345-29-5 |
| Color | Off-White |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H23NO3 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : ≥ 100 mg/mL (307.31 mM) |
| SMILES | O=C(C1=CC=C2N=C3C(CCCO3)=CC2=C1)[C@H]4CC[C@@H](OC)CC4 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?