missing translation for 'onlineSavingsMsg'
Learn More

JNJ-47117096 hydrochloride, MedChemExpress

Product Code. 16495148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495148 10 mg 10mg
16495856 100 mg 100mg
16493296 25 mg 25mg
16471377 50 mg 50mg
16427877 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495148 Supplier Medchem Express Supplier No. HY12420
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress JNJ-47117096 hydrochloride is potent and selective MELK inhibitor, with an IC50 of 23 nM, also effectively inhibits Flt3, with an IC50 of 18 nM.

JNJ-47117096 hydrochloride is potent and selective MELK inhibitor, with an IC50 of 23 nM, also effectively inhibits Flt3, with an IC50 of 18 nM.

Specifications

Chemical Name or Material JNJ-47117096 hydrochloride
CAS 1610536-69-0
Color White
Purity Grade Notes Research
Molecular Formula C21H23ClN4O2
Quantity 10 mg
Synonym MELK-T1 hydrochloride
Solubility Information DMSO : ≥ 250 mg/mL (626.74 mM) ∣H2O : 3.33 mg/mL (8.35 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC1=CC=C2CCNCCC2=C1)C3=CC=C(C4=CNN=C4)C=C3OC.Cl
Molecular Weight (g/mol) 398.89
Formula Weight 398.89
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.