Learn More
JNJ-38877618, MedChemExpress
Product Code. 16487117
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487117
Brand: Medchem Express HY111050
MedChemExpress JNJ-38877618 is a potent, highly selective, orally bioavailable Met kinase inhibitor with IC50s of 2 and 3 nM for wild type and mutant Met, respectively.
JNJ-38877618 is a potent, highly selective, orally bioavailable Met kinase inhibitor with IC50s of 2 and 3 nM for wild type and mutant Met, respectively.
Specifications
Chemical Name or Material JNJ-38877618
Physical Form Solid
CAS 943540-74-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H12F2N6
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 5 mg/mL (13.36 mM; Need ultrasonic)
SMILES FC(C1=CC=C2N=CC=CC2=C1)(F)C3=NN=C4C=CC(C5=CC=NC=C5)=NN43
Molecular Weight (g/mol) 374.35
Formula Weight 374.35
Percent Purity 98.26%
Grade Research