missing translation for 'onlineSavingsMsg'
Learn More

JNJ 16259685, TRC

Product Code. 17910725
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17910725 25 mg 25mg
17921055 10 mg 10mg
17972385 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17910725 Supplier Toronto Research Chemicals Supplier No. TRCJ21130525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409345-29-5
Molecular Formula C20 H23 N O3
Molecular Weight (g/mol) 325.4
Synonym JNJ 16259685, (3,4-Dihydro-2H-pyrano[2,3-b]quinolin-7-yl)(cis-4-methoxycyclohexyl)-methanone
IUPAC Name 3,4-dihydro-2H-pyrano[2,3-b]quinolin-7-yl-(4-methoxycyclohexyl)methanone
SMILES CO[C@@H]1CC[C@@H](CC1)C(=O)c2ccc3nc4OCCCc4cc3c2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H23NO3/c1-23-17-7-4-13(5-8-17)19(22)14-6-9-18-16(11-14)12-15-3-2-10-24-20(15)21-18/h6,9,11-13,17H,2-5,7-8,10H2,1H3/t13-,17+
Formula Weight 325.1678
Chemical Name or Material 3,4-dihydro-2H-pyrano[2,3-b]quinolin-7-yl-(4-methoxycyclohexyl)methanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.