Learn More
JH-VIII-157-02, MedChemExpress
Product Code. 16485426
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485426 100 mg 100mg
16483136 25 mg 25mg
16480877 50 mg 50mg
16464578 10 mg 10mg
16417297 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16485426 Supplier Medchem Express Supplier No. HY112140
 Restricted Item
Estimated Shipment: 16-11-2026
MedChemExpress JH-VIII-157-02 is a structural analogue of alectinib, acts as an ALK inhibitor, and shows an IC50 of 2 nM for echinoderm microtubule-associated protein-like 4-ALK (EML4-ALK) G1202R in cells.
JH-VIII-157-02 is a structural analogue of alectinib, acts as an ALK inhibitor, and shows an IC50 of 2 nM for echinoderm microtubule-associated protein-like 4-ALK (EML4-ALK) G1202R in cells.
Specifications
Chemical Name or Material JH-VIII-157-02
Physical Form Solid
CAS 1639422-97-1
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C28H27N5O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 25 mg/mL (53.70 mM)
SMILES O=C(N(C)C)CN1N=CC(C2=C(CC)C=C3C(C(C)(C)C(NC4=C5C=CC(C#N)=C4)=C5C3=O)=C2)=C1
Molecular Weight (g/mol) 465.55
Formula Weight 465.55
Percent Purity 99.67%
Grade Research