Learn More
Cole-Parmer™ Jenway™ Propane Regulator
Product Code. 11435069
Change view
Click to view available options
For Use With (Equipment):
Model FGA-500-010E
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11435069 Model FGA-500-010E Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11435069 Supplier Cole-Parmer™ Supplier No. 1265596
  • ONLINE EXCLUSIVE PRICE
    472.00 EUR
    360.44 EUR / Pack of 1
    Save 111.56 EUR
    23% Off
Estimated Shipment: 25-01-2027
Parts and accessories that keep your flame photometer operating
Specifications
Delivery Pressure Yes
Type Propane Regulator
For Use With (Equipment) Model FGA-500-010E
Gases Measured Propane
Includes 2 Stage adjustable pressure