Learn More
Jenway™ Power Supply
Product Code. 12843673
Change view
Click to view available options
For Use With (Equipment):
73 Series Spectrophotometers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12843673 73 Series Spectrophotometers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12843673 Supplier Jenway™ Supplier No. 9995056
  • ONLINE EXCLUSIVE PRICE
    169.00 EUR
    154.14 EUR / Pack of 1
    Save 14.86 EUR
    8% Off
Estimated Shipment: 02-12-2026
Plug type: UK plug
Specifications
Product Type Power Supply
For Use With (Equipment) 73 Series Spectrophotometers
Voltage 24 V
Electrical Requirements 24 V, 65 W