Learn More
Jenway™ Conductivity Standard 1413μS
Product Code. 11480499
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480499 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11480499 Supplier Jenway Supplier No. 1615659
  • ONLINE EXCLUSIVE PRICE
    90.00 EUR
    71.26 EUR / 500mL
    Save 18.74 EUR
    20% Off
View all promotions
 Restricted Item
Estimated Shipment: 21-01-2027