Learn More
GPE Scientific™ J Young Vac Line Bubbler
Product Code. 15894804
Change view
Click to view available options
Capacity (Metric):
20 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15894804 20 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15894804 Supplier GPE Scientific™ Supplier No. JYB/1
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 29-12-2026
GPE Scientific™ J Young Vac Line Bubbler is specially designed for use with Schlenk lines and vacuum manifolds.
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
Specifications
Capacity (Metric) 20 mL
Length (Metric) 130 mm
Product Type Vacuum line bubbler
Diameter (Metric) Outer 30 mm