Learn More
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Glass Key and Socket
Product Code. 15894894
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15894894 250 mL Each
15884894 25 mL Each
15804904 50 mL Each
15864894 100 mL Each
15814904 500 mL Each
15874894 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15894894 Supplier GPE Scientific™ Supplier No. SRG/250
Estimated Shipment: 23-02-2027
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Glass Key and Socket is specially designed for use with Schlenk lines and vacuum manifolds, and is available in different capacities.
  • Includes glass key and socket
  • Available in various capacities
  • Available with different joint sizes
Specifications
Capacity (Metric) 250 mL
Includes Glass key and socket
Product Type Schlenk Flask
Socket Joint Joint size: 24
Bottom Shape Round