missing translation for 'onlineSavingsMsg'
Learn More

Ivermectin, ≥95%, MP Biomedicals™

Product Code. p-4521065 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402651 250 mg 250mg
11412651 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11402651 Supplier MP Biomedicals Supplier No. 0219600983
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ivermectin is a broad-spectrum antiparasitic agent. Ivermectin is a macrolide antiobiotic, & semisynthetic derivative of avermectin B1a and B1b

Chemical Identifiers

CAS 70288-86-7
Molecular Formula C48H74O14
Molecular Weight (g/mol) 875.106
MDL Number MFCD00869511
InChI Key AZSNMRSAGSSBNP-RDKIRRGNSA-N
Synonym 22, 23-Dihydroavermectin B1, Hyvermectin
PubChem CID 122172888
SMILES CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C

Specifications

Type Ivermectin
Physical Form Crystalline Powder
Color White or White/Yellow
Quantity 250 mg
Assay Percent Range ≥95%
Formula Weight 875.1
Chemical Name or Material Ivermectin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.