missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ivacaftor
Description
Ivacaftor is a potent and selective potentiator of Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) that targets F508del-CFTR and G551D-CFTR (EC50 values are 25 nM and 100 nM, respectively); its potentiation activity depends on the level of CFTR phosphorylation. Ivacaftor reversibly binds to wild type (WT-CFTR) and G551D-CFTR mutants, increasing the open probability via an ATP-independent mechanism, and enhances cAMP/PKA-signaling mediated gating activity for both WT-CFTR and G551D-CFTR mutants. In cultured human CF bronchial epithelia with G551D/F508del mutations, Ivacaftor increases Forskolin (Cat. No. 1099)-induced transepithelial current (EC50 = 236 nM) and increases chloride secretion by 10-fold. Also rescues endothelial CFTR expression and function and prevents endothelial barrier failure and protein leak in human pulmonary microvascular endothelial cells. Ivacaftor is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-[2,4-Bis(1,1-dimethylethyl)-5-hydroxyphenyl]-1,4-dihydro-4-oxo-3-quinolinecarboxamide |
| CAS | 873054-44-5 |
| Quantity | 10 mg |
| Target | CFTR |
| Molecular Formula | C24H28N2O3 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?