Learn More
Isoxepac, TRC
Specifications
Specifications
| Chemical Name or Material | (11-oxo-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic acid |
| CAS | 55453-87-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 H12 O4 |
| Quantity | 1 g |
| Synonym | 6,11-Dihydro-11-oxodibenz[b,e]oxepin-2-acetic acid, 11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-acetic acid, 11-Oxo-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid, 11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-acetic acid, 6,11-Dihydro-11-oxodibenz[b,e]oxepin-2-acetic acid, Artil, HP 549, Isoxepac, NSC 300907, P 720549, Dibenz[b,e]oxepin-2-acetic acid, 6,11-dihydro-11-oxo- |
| InChI Formula | InChI=1S/C16H12O4/c17-15(18)8-10-5-6-14-13(7-10)16(19)12-4-2-1-3-11(12)9-20-14/h1-7H,8-9H2,(H,17,18) |
| SMILES | OC(=O)Cc1ccc2OCc3ccccc3C(=O)c2c1 |
| IUPAC Name | 2-(11-oxo-6H-benzo[c][1]benzoxepin-2-yl)acetic acid |
| Molecular Weight (g/mol) | 268.26 |
| Show More |