Learn More
Isoquercitroside, TRC
Product Code. 30136643
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136643 250 mg 250mg
30004592 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136643 Supplier Toronto Research Chemicals Supplier No. TRCI874675250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21637-25-2
Molecular Formula C21 H20 O12
Molecular Weight (g/mol) 464.38
Synonym 2-(3,4-Dihydroxyphenyl)-3-(β-D-glucofuranosyloxy)-5,7-dihydroxy-4H-1-benzopyran-4-one, 3,3',4',5,7-Pentahydroxyflavone 3-β-D-glucofuranoside, Isoquercitrin, Isoquercitroside, Isotrifoliin, Quercetin 3-(β-D-glucofuranoside), Quercetin 3-O-β-D-glucofuranoside
IUPAC Name 3-[(2S,4R,5R)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-yl]oxy-2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one
SMILES OC[C@@H](O)[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)C(O)[C@H]1O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C21H20O12/c22-6-12(27)19-16(29)17(30)21(32-19)33-20-15(28)14-11(26)4-8(23)5-13(14)31-18(20)7-1-2-9(24)10(25)3-7/h1-5,12,16-17,19,21-27,29-30H,6H2/t12-,16-,17?,19-,21+/m1/s1
Formula Weight 464.0955
Percent Purity >95%
Chemical Name or Material Isoquercitroside
Grade HPLC