Learn More
Isopyrazam, TRC
Product Code. 30134720
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134720 50 mg 50mg
30004508 5 mg 5mg
30003345 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134720 Supplier Toronto Research Chemicals Supplier No. TRCI87460050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 881685-58-1
Molecular Formula C20 H23 F2 N3 O
Molecular Weight (g/mol) 359.41
Synonym 1H-Pyrazole-4-carboxamide, 3-(difluoromethyl)-1-methyl-N-[1,2,3,4-tetrahydro-9-(1-methylethyl)-1,4-methanonaphthalen-5-yl]-, Isopyrazam, N-[1,2,3,4-Tetrahydro-9-(1-methylethyl)-1,4-methanonaphthalen-5-yl]-3-(difluoromethyl)-1-methyl-1H-pyrazol-4-carboxamide, N-[1,2,3,4-Tetrahydro-9-(1-methylethyl)-1,4-methanonaphthalen-5-yl]-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carboxamide
IUPAC Name 3-(difluoromethyl)-1-methyl-N-(11-propan-2-yl-3-tricyclo[6.2.1.02,7]undeca-2,4,6-trienyl)pyrazole-4-carboxamide
SMILES CC(C)C1C2CCC1c3c(NC(=O)c4cn(C)nc4C(F)F)cccc23
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C20H23F2N3O/c1-10(2)16-12-7-8-13(16)17-11(12)5-4-6-15(17)23-20(26)14-9-25(3)24-18(14)19(21)22/h4-6,9-10,12-13,16,19H,7-8H2,1-3H3,(H,23,26)
Formula Weight 359.1809
Chemical Name or Material Isopyrazam