missing translation for 'onlineSavingsMsg'
Learn More

Isopropylamine Hydrochloride, TRC

Product Code. 30134918
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134918 50 mg 50mg
30133887 25 mg 25mg
30009765 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134918 Supplier Toronto Research Chemicals Supplier No. TRCI82398050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15572-56-2
Molecular Formula C3H9N . HCl
Molecular Weight (g/mol) 95.57
Synonym Monoisopropylammonium Chloride, iso-Propylamine Hydrochloride
IUPAC Name propan-2-amine;hydrochloride
SMILES Cl.CC(C)N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C3H9N.ClH/c1-3(2)4;/h3H,4H2,1-2H3;1H
Formula Weight 95.0502
Chemical Name or Material Isopropylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.