missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl nitrile, TRC

Product Code. 30392201
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Product Code. Quantity
30392201 25 g
30405296 10 g
30388846 5 g
3 options
This item is not returnable. View return policy

Product Code. 30392201

Brand: Toronto Research Chemicals TRCI86831025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-82-0
Formula Weight 69.0578
InChI Formula InChI=1S/C4H7N/c1-4(2)3-5/h4H,1-2H3
IUPAC Name 2-methylpropanenitrile
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.11
SMILES CC(C)C#N
Synonym 2-methyl-Propanenitrile, Isobutyronitrile (6CI,8CI), 2-Methylpropanenitrile, 1-Cyano-1-methylethane, 2-Cyanopropane, 2-Methylpropionitrile, Dimethylacetonitrile, Isobutanenitrile, Isopropyl cyanide, NSC 60536, alpha-Methylpropanenitrile, alpha-Methylpropionitrile
Chemical Name or Material Isopropyl nitrile

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.