accessibility menu, dialog, popup

UserName

CH3CH3N

CAS RN 78-82-0

IUPAC Name: 2-methylpropanenitrile
Synonyms: isobutyronitrile isopropyl nitrile isopropylkyanid isopropylnitrile
Molecular Weight (g/mol): 69.11
Molecular Formula: C4H7N
InChi Key: LRDFRRGEGBBSRN-UHFFFAOYSA-N
SMILES: CC(C)C#N
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)