Learn More
Isopropyl Diphenyl Phosphate-d7, TRC
Specifications
Specifications
| Chemical Name or Material | Isopropyl Diphenyl Phosphate-d7 |
| CAS | 2469626-65-9 |
| Molecular Formula | C15H10D7O4P |
| Quantity | 100 mg |
| Synonym | Diphenyl Isopropyl Phosphate-d7, 1-Methylethyl Diphenyl Ester Phosphoric Acid-d7 |
| InChI Formula | InChI=1S/C15H17O4P/c1-13(2)17-20(16,18-14-9-5-3-6-10-14)19-15-11-7-4-8-12-15/h3-13H,1-2H3/i1D3,2D3,13D |
| SMILES | O=P(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC(C([2H])([2H])[2H])([2H])C([2H])([2H])[2H] |
| IUPAC Name | diphenyl (propan-2-yl-d7) phosphate |
| Molecular Weight (g/mol) | 299.31 |
| Formula Weight | 299.13 |