missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-β-D-Thiogalactopyranoside, MP Biomedicals™

Product Code. 11348959 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348959 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11348959 Supplier MP Biomedicals Supplier No. 02102101.2
  • ONLINE EXCLUSIVE PRICE
    40.80 EUR
    38.85 EUR / 250mg
    Save 1.95 EUR
    4% Off
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Commonly used as a reagent in cloning procedures. Isopropyl-β-D-Thiogalactopyranoside, MP Biomedicals (IPTG) is an analogue of allolactose that binds specifically to the repressor protein of the lac operon and induces expression of β-galactosida

IPTG is a commonly used reagent in cloning procedures that require induction of b-galactosidase activity and is used in conjunction with X-Gal in blue-white color selection of recombinant bacterial colonies.

Specifications

CAS 367-93-1
Melting Point 105°C to 115°C
pH 5.0 to 7.0
Molecular Formula C9H18O5S
MDL Number MFCD00063273
Quantity 250 mg
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
Solubility Information Soluble in methanol and water (50mg/mL, clear, colorless solution).
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Optical Rotation −26° to −33° (c=1, water)
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 238.30
PubChem CID 656894
ChEBI CHEBI:61448
Formula Weight 238.3
Percent Purity ≥99%
Grade Not Available
Concentration or Composition (by Analyte or Components) Working Concentration: The final concentration typically used in indicator plates is 0.2 mM.
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.