missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Acetate, Sigma-Aldrich™

Product Code. 15881031
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15881031 2.5 L 2.5 L
15846650 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15881031 Supplier Sigma Aldrich Supplier No. 8205602500
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biorenewable and alternative to petrochemical solvents. Ethanol is environmentally preferable green solvent and thus it aligns with Safer Solvents and Auxiliaries and Use of Renewable Feedstocks.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-21-4

Specifications

Melting Point -73°C
Density 0.87 g/cm3 at 20°C
Boiling Point 88°C to 90°C/1013 hPa
Molecular Formula CH3COOCH(CH3)2
MDL Number MFCD00008877
Quantity 2.5 L
Synonym Isopropyl acetate, Isopropyl acetate
Molecular Weight (g/mol) 102.13
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.