missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 2-Furoate (~90%), TRC

Product Code. 30179098
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179098 2.5 g 2.5g
30178949 500 mg 500mg
30178974 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179098 Supplier Toronto Research Chemicals Supplier No. TRCI8245652.5G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6270-34-4
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.16
Synonym Isopropyl 2-furylcarboxylate, NSC 35576
IUPAC Name propan-2-yl furan-2-carboxylate
SMILES CC(C)OC(=O)c1occc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C8H10O3/c1-6(2)11-8(9)7-4-3-5-10-7/h3-6H,1-2H3
Formula Weight 154.063
Chemical Name or Material Isopropyl 2-Furoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.