Learn More
Isopimpinellin, TRC
Product Code. 30006896
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006896 50 mg 50mg
18031650 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30006896 Supplier Toronto Research Chemicals Supplier No. TRCI82195050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 482-27-9
Molecular Formula C13 H10 O5
Molecular Weight (g/mol) 246.22
Synonym Isopimpinellin (6CI), 4,9-Dimethoxy-7H-furo[3,2-g][1]benzopyran-7-one, 5,8-Dimethoxy-6,7-furanocoumarin, 5,8-Dimethoxypsoralen, 5-Methoxyxanthotoxin, 8-Methoxybergapten, Isoimpinellin, NSC 217988, NSC 401288
IUPAC Name 4,9-dimethoxyfuro[3,2-g]chromen-7-one
SMILES COc1c2OC(=O)C=Cc2c(OC)c3ccoc13
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C13H10O5/c1-15-10-7-3-4-9(14)18-12(7)13(16-2)11-8(10)5-6-17-11/h3-6H,1-2H3
Formula Weight 246.0528
Chemical Name or Material Isopimpinellin