missing translation for 'onlineSavingsMsg'
Learn More

Isophorone diisocyanate, 99.3%, For GC analysis, MP Biomedicals™

Product Code. 11413312 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413312 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11413312 Supplier MP Biomedicals Supplier No. 0215510680
  • ONLINE EXCLUSIVE PRICE
    48.50 EUR
    46.20 EUR / 100g
    Save 2.30 EUR
    4% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 4098-71-9
Color Colorless
Density 1.058g/cm3 at 20°C
Molecular Formula C12H18N2O2
MDL Number MFCD00064956
Quantity 100 g
Synonym isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
InChI Key NIMLQBUJDJZYEJ-UHFFFAOYNA-N
SMILES CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
IUPAC Name 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
Molecular Weight (g/mol) 222.29
PubChem CID 169132
ChEBI CHEBI:53214
Formula Weight 222.3
Physical Form Liquid
Chemical Name or Material Isophorone diisocyanate, For GC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.