Learn More
Isopentyl 3-(methylthio)propionate, 98%, Thermo Scientific™
Product Code. 11485996
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11485996
Brand: Thermo Scientific Alfa Aesar B22313.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93762-35-7
Molecular Formula CH3SCH2CH2CO2CH2CH2CH(CH3)2
MDL Number MFCD00053101
Synonym isopentyl 3-methylthio propionate, isoamyl 3-methylthio propionate, 3-methylbutyl 3-methylsulfanyl propanoate, 3-methylbutyl 3-methylthio propanoate, 3-methylbutyl 3-methylthio propionate
Specifications
Density 0.976
Boiling Point 222°C
Flash Point 115°C (239°F)
Refractive Index 1.4585
Quantity 100 g
Beilstein 7914388
Formula Weight 190.31
Percent Purity 98%
Chemical Name or Material Isopentyl 3-(methylthio)propionate