accessibility menu, dialog, popup

UserName

CAS RN 93762-35-7

H3COOSCH3

CAS RN 93762-35-7

IUPAC Name: pentyl 3-(methylsulfanyl)propanoate
Synonyms: pentyl 3-(methylsulfanyl)propanoate
Molecular Weight (g/mol): 190.3
Molecular Formula: C9H18O2S
InChi Key: VSCXQPTWKZWPQB-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)CCSC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results