missing translation for 'onlineSavingsMsg'
Learn More

Isometronidazole, TRC

Product Code. 30012195
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012195 1 g 1g
30004559 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012195 Supplier Toronto Research Chemicals Supplier No. TRCI8213401G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 705-19-1
Molecular Formula C6 H9 N3 O3
Molecular Weight (g/mol) 171.15
Synonym 2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethanol, Metronidazole Imp. E (EP), 2-Methyl-4-nitro-1H-imidazole-1-ethanol, 2-Methyl-4-nitroimidazole-1-ethanol, 1-(2-Hydroxyethyl)-2-methyl-4-nitroimidazole, Isometronidazole, Izoklion, Mizonidazole, RP 8979, SC-16427
IUPAC Name 2-(2-methyl-4-nitroimidazol-1-yl)ethanol
SMILES Cc1nc(cn1CCO)[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H9N3O3/c1-5-7-6(9(11)12)4-8(5)2-3-10/h4,10H,2-3H2,1H3
Formula Weight 171.0644
Percent Purity >95%
Chemical Name or Material 2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.