Learn More
Isometronidazole-d4 (hydroxyethyl-d4), CDN
Specifications
Specifications
| Chemical Name or Material | Isometronidazole-D4 |
| CAS | 2469037-64-5 |
| Molecular Formula | C6 D4 H5 N3 O3 |
| Quantity | 0.005 g |
| Synonym | 2-(2-Methyl-4-nitro-1 H-imidazol-1-yl)ethanol-1,1,2,2-D4, Isometronidazole-d4 (hydroxyethyl-d4), 2-Methyl-4-nitroimidazole-1-ethanol-1,1,2,2-d4, 1-(2-Hydroxyethyl-d4)-2-methyl-4-nitroimidazole |
| InChI Formula | InChI=1 S/C6H9N3O3/c1-5-7-6(9(11)12)4-8(5)2-3-10/h4,10 H,2-3H2,1H3/i2D2,3D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])n1cc(nc1C)[N+](=O)[O-] |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-(2-methyl-4-nitroimidazol-1-yl)ethanol |
| Molecular Weight (g/mol) | 175.1786 |
| Formula Weight | 175.0895 g/mol |
| Show More |