Learn More
Isomangiferin, MedChemExpress
Product Code. 16491498
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491498 5 mg 5mg
16489088 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16491498 Supplier Medchem Express Supplier No. HYN0772
MedChemExpress Isomangiferin, a natural product, is reported to have antiviral activity.
Isomangiferin, a natural product, is reported to have antiviral activity.
Specifications
Chemical Name or Material Isomangiferin
Physical Form Solid
CAS 24699-16-9
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H18O11
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 50 mg/mL (118.39 mM; Need ultrasonic)
SMILES OC1=CC(O)=C2C(OC3=CC(O)=C(O)C=C3C2=O)=C1[C@@H]([C@@H]([C@@H](O)[C@@H]4O)O)O[C@@H]4CO
Molecular Weight (g/mol) 422.34
Formula Weight 422.34
Percent Purity 99.6%
Grade Research