missing translation for 'onlineSavingsMsg'
Learn More

Isochlorogenic acid A, MedChemExpress

Product Code. 16474737
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474737 50 mg 50mg
16458848 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16474737 Supplier Medchem Express Supplier No. HYN0056
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Isochlorogenic acid A (3,5-Dicaffeoylquinic acid) is a natural phenolic acid with antioxidant and anti-inflammatory activities .

Isochlorogenic acid A (3,5-Dicaffeoylquinic acid) is a natural phenolic acid with antioxidant and anti-inflammatory activities .

Specifications

Chemical Name or Material Isochlorogenic acid A
CAS 2450-53-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H24O12
Quantity 50 mg
Synonym 3,5-Dicaffeoylquinic acid 3,5-CQA
Solubility Information DMSO : ≥ 50 mg/mL (96.81 mM)
SMILES OC(C=C1/C=C/C(O[C@H]2[C@@H]([C@@H](C[C@](O)(C2)C(O)=O)OC(/C=C/C3=CC(O)=C(C=C3)O)=O)O)=O)=C(C=C1)O
Molecular Weight (g/mol) 516.45
Formula Weight 516.45
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.