missing translation for 'onlineSavingsMsg'
Learn More

Isobutyraldehyde, Sigma-Aldrich™

Product Code. 15858590
Change view
Click to view available options
Quantity:
100 mL
1 L
20 kg
Unit Size:
1 L
100mL
20kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15858590 20 kg 20kg
15838590 100 mL 100mL
15848590 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15858590 Supplier Sigma Aldrich Supplier No. 8015569020
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-84-2

Specifications

Melting Point -65.9°C
Density 0.79 g/cm3 at 20°C
Boiling Point 63°C to 64.5°C/1013 mbar
Molecular Formula (CH3)2CHCHO
MDL Number MFCD00006980
Quantity 20 kg
Synonym Isobutyraldehyde, Isobutyraldehyde
Molecular Weight (g/mol) 72.11
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.