Learn More
Isoamyl Propionate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 105-68-0 |
| Melting Point | -73°C |
| Density | 0.869 g/cm3 at 20°C |
| Molecular Formula | (CH3)2CHCH2CH2OCOCH2CH2CH3 |
| MDL Number | MFCD00048611 |
| Quantity | 100 mL |
| Synonym | Isoamyl propionate, Propionic acid isoamyl ester, Isoamyl propionate |
| Molecular Weight (g/mol) | 144.21 |
| Grade | Research |