missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl alcohol, Supelco™

Código de producto. 32237590
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
4 L
missing translation for 'unitSize':
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity missing translation for 'unitSize'
32237590 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Código de producto. 32237590 missing translation for 'mfr' Supelco missing translation for 'supplierNo' 1009794000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For analysis EMSURE™ ACS, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoamyl alcohol
CAS 123-51-3
Melting Point -117°C (lit.)
Density 0.809 g/mL at 25°C (lit.)
Boiling Point 130°C (lit.)
Flash Point 43.5°C (closed cup)
Molecular Formula (CH3)2CHCH2CH2OH
Quantity 4 L
Synonym 3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol, 3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol
Molecular Weight (g/mol) 88.15
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.