missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Alcohol, Sigma-Aldrich™

Product Code. 15874471
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874471 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15874471 Supplier Sigma Aldrich Supplier No. 8189691000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 123-51-3
Melting Point −117°C (Lit.)
Density 0.809 g/mL at 25°C
Boiling Point 130°C
Molecular Formula (CH3)2CHCH2CH2OH
MDL Number MFCD00002934
Quantity 1 L
Synonym 3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol, 3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol
Molecular Weight (g/mol) 88.15
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.