Learn More
Brady™ ISO Square Polyester Safety Sign 'Fire Alarm Flashing Light'
Product Code. 17841795
Change view
Click to view available options
Dimensions (L x W):
100 x 100 mm
148 x 148 mm
150 x 150 mm
200 x 200 mm
250 x 250 mm
315 x 315 mm
400 x 400 mm
Color:
Photoluminescent on Red
White on Red
Unit Size:
Each
Certifications/Compliance:
Class B
Class C
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Color unitSize Certifications/Compliance
17841795 100 x 100 mm White on Red Each -
17891765 148 x 148 mm White on Red Each -
17871765 200 x 200 mm White on Red Each -
17881765 250 x 250 mm White on Red Each -
17821695 315 x 315 mm White on Red Each -
17811775 400 x 400 mm White on Red Each -
17861695 100 x 100 mm Photoluminescent on Red Each Class B
17861715 100 x 100 mm Photoluminescent on Red Each Class C
17851795 150 x 150 mm Photoluminescent on Red Each Class B
17801705 150 x 150 mm Photoluminescent on Red Each Class C
17801775 200 x 200 mm Photoluminescent on Red Each Class B
17851775 200 x 200 mm Photoluminescent on Red Each Class C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 17841795 Supplier Brady™ Supplier No. 311199
  • ONLINE EXCLUSIVE PRICE
    3.25 EUR
    3.18 EUR / Pack of 1
    Save 0.07 EUR
    2% Off
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
ISO Safety Sign displaying flashing fire alarm and flames.
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Alternate Part Number F/F018/NT/PE-100X100-1
Finish Gloss
Material Polyester
Quantity 1 Sign
Adhesive Type Permanent
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 3.94 in.
Width (English) 3.94 in.
Temperature Range (Metric) -40°C to +110°C
Shape Square
Temperature Range (English) -40°F to +230°F
Languages Universal
Dimensions (L x W) 100 x 100 mm
Displayed Text Fire Alarm Flashing Light
Product Type Safety Sign
Color White on Red