Learn More
Brady™ ISO Safety Sign without Text - Warning: Hot Surface
Product Code. 15629673
Change view
Click to view available options
Dimensions (L x W):
9 x 10 mm
22 x 25 mm
Unit Size:
Each
Pack of 54
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15629673 9 x 10 mm Pack of 54
15963057 22 x 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15629673 Supplier Brady™ Supplier No. 139013
  • ONLINE EXCLUSIVE PRICE
    11.40 EUR
    10.94 EUR / Pack of 54
    Save 0.46 EUR
    4% Off
View all promotions
In Stock
Add to basket
Add to basket
Safety sign for hazard identification.
  • Sign compliant to ISO 7010
  • Improves workplace safety
Specifications
Alternate Part Number W/W017/NT-SA-TRI 10/54-B
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Shape Triangular
Dimensions (L x W) 9 x 10 mm
Color Black on Yellow