missing translation for 'onlineSavingsMsg'
Learn More
Learn More
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
30280933
|
Thermo Scientific
T004610500 |
57.55 EUR 46.40 EUR / 500mL Save 11.15 EUR 19% Off |
Estimated Shipment: 20-11-2026
Log in to see stock. |
||||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
30280934
|
Thermo Scientific
T004611000 |
87.35 EUR 84.80 EUR / 1 L Save 2.55 EUR 2% Off |
In Stock
Log in to see stock. |
||||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
30280935
|
Thermo Scientific
T004612500 |
254.00 EUR 179.00 EUR / 2.5 L Save 75.00 EUR 29% Off |
In Stock
Log in to see stock. |
||||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Isopropanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contain: Isopropanol
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.