Learn More
Brady™ ISO 7010 Sign - Warning, Electricity
Product Code. 15684717 Shop All Brady Products
Change view
Click to view available options
Height (Metric):
87 mm
173 mm
Width (Metric):
100 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Unit Size
15684717 173 mm 200 mm Each
15674717 87 mm 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15684717 Supplier Brady™ Supplier No. 828100
  • ONLINE EXCLUSIVE PRICE
    11.20 EUR
    10.40 EUR / Pack of 1
    Save 0.80 EUR
    7% Off
View all promotions
In Stock
Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC W012-Tri 200-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 173 mm
Thickness 1.501 mm
Width (Metric) 200 mm
Temperature Range (Metric) -40°C to 120°C
Shape Triangular
Languages Global
Color Black on Yellow