Learn More
Brady™ ISO 7010 Sign - Fire alarm call point
Product Code. 15684407
Change view
Click to view available options
Height (Metric):
148 mm
200 mm
Width (Metric):
148 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) unitSize
15684407 148 mm 148 mm Each
15694407 200 mm 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15684407 Supplier Brady™ Supplier No. 817682
  • ONLINE EXCLUSIVE PRICE
    9.45 EUR
    8.78 EUR / Pack of 1
    Save 0.67 EUR
    7% Off
View all promotions
In Stock
Add to basket
Add to basket
Prevent costly accidents with ISO 7010 safety signs for optimal impact and recognizability.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F005-148x148-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 148 mm
Thickness 1.501 mm
Width (Metric) 148 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Languages Global
Color White on Red