missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ISG15 Activating Enzyme/UBE1L Antibody (SR4330), Novus Biologicals™
Shop All Bio Techne ProductsDescription
ISG15 Activating Enzyme/UBE1L Monoclonal antibody specifically detects ISG15 Activating Enzyme/UBE1L in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | ISG15 Activating Enzyme/UBE1L |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | SR4330 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 |
| Formulation | PBS (pH 7.4), 150mM NaCl, 50% glycerol |
| Gene Alias | D8UBA1, ubiquitin-activating enzyme E1 homolog B, UBA1B, UBA7, ubiquitin-activating enzyme E1, UBE1L, UBE2MGC12713, Ubiquitin-activating enzyme 7, Ubiquitin-activating enzyme E1 homolog, ubiquitin-activating enzyme E1-like, ubiquitin-activating enzyme E1-related protein, ubiquitin-activating enzyme-2, ubiquitin-like modifier activating enzyme 7, ubiquitin-like modifier-activating enzyme 7 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human ISG15 Activating Enzyme/UBE1L (Uniprot #: P41226) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?