Learn More
Mettler Toledo™ ISA Aluminum Sulfate Solution
Product Code. 12659255
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12659255 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12659255 Supplier Mettler Toledo™ Supplier No. 51350108
  • ONLINE EXCLUSIVE PRICE
    105.00 EUR
    79.84 EUR / 250mL
    Save 25.16 EUR
    23% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-12-2026
Specifications
Description ISA solution 0.9 mol/L Al2(SO4)3, 250ml
Type ISA Aluminum Sulfate Solution
Quantity 250 mL
compliance-icons
Product Identifier
  • ISA solution Al sulfate 0.9 M
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Aluminium sulphate