Learn More
Micronova™ Irradiated Sharpie Fine Point Marker
Product Code. 13167726 Shop All Micronova Products
Change view
Click to view available options
Color:
Black
Blue
Red
Length (English):
6 in.
7 in.
Packaging:
Individually bagged, 4 per double bag, 12 per third bag (case pack)
Individually double bagged, 12 per outer bag (case pack)
Unit Size:
Pack of 12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (English) Packaging Unit Size
13167726 Red 6 in. Individually bagged, 4 per double bag, 12 per third bag (case pack) Pack of 12
12837053 Black 6 in. Individually bagged, 4 per double bag, 12 per third bag (case pack) Pack of 12
15387863 Black 6 in. Individually double bagged, 12 per outer bag (case pack) Pack of 12
13177726 Black 7 in. Individually bagged, 4 per double bag, 12 per third bag (case pack) Pack of 12
13157726 Blue 6 in. Individually bagged, 4 per double bag, 12 per third bag (case pack) Pack of 12
13197726 Blue 7 in. Individually bagged, 4 per double bag, 12 per third bag (case pack) Pack of 12
13107736 Red 7 in. Individually bagged, 4 per double bag, 12 per third bag (case pack) Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 13167726 Supplier Micronova™ Supplier No. PEN23IR
  • ONLINE EXCLUSIVE PRICE
    49.90 EUR
    46.24 EUR / Pack of 12
    Save 3.66 EUR
    7% Off
 Restricted Item
Estimated Shipment: 05-02-2027
Irradiated, fine point permanent marker with fade and water resistant, quick drying, non-toxic ink for use in aseptic environments. Available capped or retractable styles.
Fade and water resistant ink
Micronova Irradiated Sharpie Fine Point Marker is the original pen style, industry standard permanent marker. This cleanroom sharpie marks permanently on most surfaces with quick drying, non-toxic, alcohol based ink that is fade and water resistant to further extend multiple project uses.
  • Fine point sharpie marker
  • Marks permanently on most surfaces
  • Writes on cold or wet surfaces, plastic bags and disposable lab-ware
  • Nontoxic, permanent, quick drying, alcohol-based ink
  • Fade and water resistant
  • Available capped or retractable styles, red, blue or black inks
  • Gamma irradiated for Aseptic ISO 5 and other clean production areas
Specifications
Material HDPE
Color Red
Cover Material HDPE
Description Retractable fine tip permanent marker
Dimensions (L x W x H) 6 x 0.5 x 0.5 in. (15.24 x 1.27 x 1.27 cm)
Length (English) 6 in.
Length (Metric) 15.2 cm
Width (English) 0.5 in.
Width (Metric) 1.27 cm
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 Certified Facility; Certificate of Irradiation Processing; Certificate of Conformance
Diameter (English) Outer 0.5 in.
Diameter (Metric) Outer 1.27 cm
Disposable Reusable
For Use With (Application) Cleanroom and Aseptic Environment Documentation and Labeling
Includes Cap
Packaging Individually bagged, 4 per double bag, 12 per third bag (case pack)
Product Type Cleanroom Marker
Quantity 12/Pk.
Size 8 x 8 x 8 in.
Sterility Sterile
Tip Style 1.0 mm Fine Point