Learn More
Bochem™ Iron Wire Flask Brushes
Product Code. 10535991
Change view
Click to view available options
Length (Metric):
420 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10535991 420 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10535991 Supplier Bochem™ Supplier No. 12341
  • ONLINE EXCLUSIVE PRICE
    7.30 EUR
    5.73 EUR / Pack of 1
    Save 1.57 EUR
    21% Off
Estimated Shipment: 17-12-2026
With natural bristles. BOCHEM™ Iron Wire Flask Brushes make cleaning flasks quick and easy.
  • Iron wire, zincked
  • Natural bristles
Specifications
Length (Metric) 420 mm
Length (Metric) Brush 140 mm