missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Chloride, Sigma-Aldrich™

Product Code. 15896370
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896370 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15896370 Supplier Sigma Aldrich Supplier No. 8039451000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

Anhydrous for synthesis

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 306°C (Decomposition)
CAS 7705-08-0
Quantity 1 kg
Density 2.89 g/cm3 at 25°C
MDL Number MFCD00011005
Synonym Iron(III) chloride, Ferric chloride, Iron trichloride, Iron(III) chloride
Molecular Weight (g/mol) 162.2
Grade Research
Chemical Name or Material Iron(III) Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.