Learn More
Iron(III) Chloride Hexahydrate, Supelco™
Product Code. 15867200
Change view
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15867200 25 kg 25kg
15824380 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15867200 Supplier Supelco Supplier No. 1039439025
 Restricted Item
Estimated Shipment: 19-11-2026
For analysis EMSURE™ ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10025-77-1
Specifications
Boiling Point 280°C to 285°C
pH 1.8 (25°C, 10 g/L in H2O)
Quantity 25 kg
Molecular Formula FeCl3 · 6H2O
MDL Number MFCD00149712
Synonym Ferric chloride hexahydrate, Iron trichloride, Ferric chloride hexahydrate, Ferric chloride hexahydrate
Molecular Weight (g/mol) 270.3
Percent Purity 99.0 to 102.0% FeCl3 basis (iodometric)
Grade Research
Chemical Name or Material Iron(III) Chloride Hexahydrate