missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride hexahydrate, MilliporeSigma™

Product Code. 15814380
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15814380 250 g Plastic Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15814380 Supplier Supelco Supplier No. 1039430250
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Specifications

CAS 10025-77-1
Quantity 250 g
Molecular Formula Cl3FeH12O6
MDL Number MFCD00149712
Synonym Ferric chloride hexahydrate; Iron trichloride
Solubility Information Soluble in water: 920g/L at 10°C
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
IUPAC Name iron(3+) hexahydrate trichloride
Molecular Weight (g/mol) 270.29
Formula Weight 270.33
Percent Purity 99.0 to 102.0%
Grade ACS
Odor Chlorine
Packaging Plastic Bottle
Chemical Name or Material Iron(III) chloride hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.