Learn More
Iron(II) Sulfate Heptahydrate, Supelco™
Description
We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives. IRON(II) SULFATE HEPTAHYDRATE is essential for developing greener energy solutions, particularly in iron-based lithium-ion batteries. Its eco-friendly properties enhance energy storage efficiency, reduce reliance on harmful materials, and support the advancement of sustainable technologies.
Specifications
Specifications
| Melting Point | >60°C (Elimination of water of crystallisation) |
| Quantity | 25 kg |
| MDL Number | MFCD00149719 |
| Synonym | Iron(II) sulfate heptahydrate, Iron, Iron(II) sulfate heptahydrate |
| Molecular Weight (g/mol) | 278.01 |
| Grade | Research |
| Density | 1.89 g/cm3 at 20°C |
| Chemical Name or Material | Iron(II) Sulfate Heptahydrate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.