Learn More
Iron(II) Chloride Tetrahydrate, Supelco™
Product Code. 15847200
Change view
Click to view available options
Quantity:
1 kg
50 kg
Unit Size:
1kg
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15847200 50 kg 50kg
15837200 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15847200 Supplier Supelco Supplier No. 1038619050
 Restricted Item
Estimated Shipment: 19-11-2026
For analysis EMSURE™
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13478-10-9
Tekniske data
Melting Point 105°C to 110°C (Elimination of water of crystallisation)
Quantity 50 kg
MDL Number MFCD00149709
Synonym Iron(II) chloride tetrahydrate, Iron dichloride, Iron(II) chloride tetrahydrate
Molecular Weight (g/mol) 198.81
Grade Research
Density 1.93 g/cm3 at 20°C
Chemical Name or Material Iron(II) Chloride Tetrahydrate
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.